This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03974839_g1
See other A630072L19RIK GE Assays ›
Gene Symbol:
A630072L19Rik
Gene Name:
RIKEN cDNA A630072L19 gene
Gene Aliases:
ENSMUSG00000072877
Chromosome Location:
Chr.12: 100939133 - 100944220 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03974839_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A630072L19Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK089356.1 - 2 - 3 649 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream G protein-coupled receptor 68 Gpr68 BB131428,
Ogr1
-34.5kb Anti-sense Chr. 12 - 100876682 - 100908198 on NCBI Build GRCm38
Downstream predicted gene,
36756
Gm36756 41.5kb Sense Chr. 12 - 100984777 - 101026968 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Probe spans exons

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 303957 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0