This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03967264_g1
See other TOMM6OS GE Assays ›
Gene Symbol:
Tomm6os
Gene Name:
translocase of outer mitochondrial membrane 6, opposite strand
Gene Aliases:
Gm14872, OTTMUSG00000018355, Prickle4os
Chromosome Location:
Chr.17: 47687610 - 47691092 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03967264_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tomm6os
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045945.1 1 - 2 72
GenBank mRNA AK046990.1 - 1 - 2 507 72

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream translocase of outer mitochondrial membrane 6 homolog (yeast) Tomm6 1110002E23Rik,
AI663970
Anti-sense Chr. 17 - 47686645 - 47688386 on NCBI Build GRCm38
Downstream fibroblast growth factor receptor substrate 3,
opposite strand
Frs3os 4930417B13Rik,
Gm14873,
OTTMUSG00000018358
11.3kb Anti-sense Chr. 17 - 47699969 - 47701996 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002792.mm.1

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700008879.mm.1 NR_045945.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008879.mm.1 NR_045945.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008879.mm.1 TC1700002792.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700008877.mm.1 NR_045945.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008877.mm.1 NR_045945.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700008877.mm.1 TC1700002792.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0