This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03957434_m1
See other 4930591A17RIK GE Assays ›
Gene Symbol:
4930591A17Rik
Gene Name:
RIKEN cDNA 4930591A17 gene
Gene Aliases:
-
Chromosome Location:
Chr.2: 179414936 - 179416880 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03957434_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930591A17Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026596.2 NP_080872.1 1 - 2 163 91
GenBank mRNA AK019836.1 - 1 - 2 156 91
AK019837.1 - 1 - 2 166 91
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY716028.1 - 1 - 2 153 Chr. 2 - 179414946 - 179416775 [+] on Build GRCm38 91
GenBankmRNA AV465038.1 - 1 - 2 139 Chr. 2 - 179414958 - 179416574 [+] on Build GRCm38 91
GenBankmRNA BY716034.1 - 1 - 2 166 Chr. 2 - 179414936 - 179416765 [+] on Build GRCm38 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
35695
Gm35695 -749.1kb Sense Chr. 2 - 178659345 - 178665836 on NCBI Build GRCm38
Downstream cadherin 4 Cdh4 AW120700,
R-Cadh,
R-cadherin,
Rcad
26.6kb Sense Chr. 2 - 179442478 - 179899377 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK019836.1, AK019837.1, NM_026596.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene 4930591A17Rik

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200022605.mm.1 NM_026596.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022605.mm.1 NM_026596.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200022605.mm.1 TC0200002771.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200022606.mm.1 NM_026596.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022606.mm.1 NM_026596.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200022606.mm.1 TC0200002771.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0