This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956497_s1
See other CRKL GE Assays ›
Gene Symbol:
Crkl
Gene Name:
v-crk sarcoma virus CT10 oncogene homolog (avian)-like
Gene Aliases:
1110025F07Rik, AA589403, AI325100, Crkol
Chromosome Location:
Chr.16: 17451985 - 17487440 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956497_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crkl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001277231.1 NP_001264160.1 2 3949 103
NM_007764.5 NP_031790.2 3 4415 103
GenBank mRNA AK003911.1 - 1 346 103
AK087977.1 - 1 1271 103
BF453354.1 - 1 169 103
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BP764489.1 - 1 Chr. 16 - 17486755 - 17487291 [-] on Build GRCm38 103
GenBankmRNA BY761703.1 - 1 364 Chr. 16 - 17486539 - 17487432 [+] on Build GRCm38 103
GenBankmRNA AA469692.1 - 1 138 Chr. 16 - 17486668 - 17487076 [+] on Build GRCm38 103
GenBankmRNA BF023362.1 - 1 412 Chr. 16 - 17486400 - 17486936 [+] on Build GRCm38 103
GenBankmRNA BY466799.1 - 1 44 Chr. 16 - 17486769 - 17487288 [+] on Build GRCm38 103
GenBankmRNA BP761857.1 - 1 Chr. 16 - 17486760 - 17487291 [-] on Build GRCm38 103
GenBankmRNA BY704166.1 - 1 346 Chr. 16 - 17486460 - 17487408 [+] on Build GRCm38 103
GenBankmRNA BQ177832.1 - 1 525 Chr. 16 - 17486281 - 17486990 [+] on Build GRCm38 103
GenBankmRNA CA876152.1 - 1 Chr. 16 - 17486744 - 17487288 [-] on Build GRCm38 102
GenBankmRNA AA589403.1 - 1 294 Chr. 16 - 17486512 - 17487034 [+] on Build GRCm38 103
GenBankmRNA AA208873.1 - 1 33 Chr. 16 - 17486773 - 17487182 [+] on Build GRCm38 104
GenBankmRNA BM235304.2 - 1 Chr. 16 - 17486764 - 17487159 [-] on Build GRCm38 103
GenBankmRNA BQ964066.1 - 1 293 Chr. 16 - 17486517 - 17487435 [+] on Build GRCm38 103
GenBankmRNA BY755012.1 - 1 77 Chr. 16 - 17486729 - 17487420 [+] on Build GRCm38 103
GenBankmRNA CB205780.1 - 1 404 Chr. 16 - 17486414 - 17487230 [+] on Build GRCm38 103
GenBankmRNA BE648808.1 - 1 111 Chr. 16 - 17486695 - 17487050 [+] on Build GRCm38 103
GenBankmRNA CK635102.1 - 1 176 Chr. 16 - 17486630 - 17487291 [+] on Build GRCm38 103
GenBankmRNA BU705483.1 - 1 178 Chr. 16 - 17486703 - 17487305 [+] on Build GRCm38 103
GenBankmRNA BP762307.1 - 1 Chr. 16 - 17486601 - 17487291 [-] on Build GRCm38 103
GenBankmRNA AA763593.1 - 1 306 Chr. 16 - 17486500 - 17487034 [+] on Build GRCm38 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream synaptosomal-associated protein 29 Snap29 1300018G05Rik,
AI891940,
AU020222,
BB131856,
Gs32
-55.6kb Sense Chr. 16 - 17406000 - 17430826 on NCBI Build GRCm38
Downstream apoptosis-inducing factor,
mitochondrion-associated 3
Aifm3 2810401C16Rik,
AI840249,
Aifl
2.2kb Sense Chr. 16 - 17489616 - 17507485 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crkl

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

blood vessel development
pattern specification process
heart development
organ morphogenesis
anterior/posterior pattern specification
thymus development
parathyroid gland development
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600001781.mm.1 NM_001277231.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001781.mm.1 NM_007764.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600001781.mm.1 TC1600000216.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600001783.mm.1 NM_001277231.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001783.mm.1 NM_007764.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600001783.mm.1 TC1600000216.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600001782.mm.1 NM_001277231.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600001782.mm.1 NM_007764.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600001782.mm.1 TC1600000216.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0