This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956427_g1
See other 1110013I04RIK GE Assays ›
Gene Symbol:
1110013I04Rik
Gene Name:
RIKEN cDNA 1110013I04 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956427_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1110013I04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK003663.1 - 1 - 2 258 77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY726277.1 - 2 - 3 267 Chr. 8 - 112895446 - 112966340 [+] on Build GRCm38 77
GenBankmRNA BY703926.1 - 1 - 2 259 Chr. 8 - 112961448 - 112962560 [+] on Build GRCm38 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream contactin associated protein-like 4 Cntnap4 Caspr4,
E130114F09Rik
-78.7kb Sense Chr. 8 - 112569965 - 112882707 on NCBI Build GRCm38
Downstream predicted gene 10280 Gm10280 ENSMUSG00000069883 104.7kb Sense Chr. 8 - 113067262 - 113093334 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent ABI Illumina

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_52_P1130599 AK003663.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105935 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176180 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176163 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176177 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07203631 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176145 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176186 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176158 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176190 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176168 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176159 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176169 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176161 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07047767 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176162 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07252687 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176150 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176184 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176199 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176155 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176174 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176193 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176196 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176165 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176179 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176148 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176189 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176191 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176147 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176164 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176170 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176171 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176152 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176188 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26176175 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06975656 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 379581 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104010152 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0