This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03956074_s1
See other CET1 GE Assays ›
Gene Symbol:
Cet1
Gene Name:
cerebellum expressed transcript 1
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03956074_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cet1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA U03988.1 - 1 451 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream kalirin,
RhoGEF kinase
Kalrn 2210407G14Rik,
AV235988,
DUET,
E530005C20Rik,
Gm539,
Hapip,
TRAD
Anti-sense Chr. 16 - 33969073 - 34573291 on NCBI Build GRCm38
Downstream ropporin,
rhophilin associated protein 1
Ropn1 1700008N21Rik,
ODF6,
RHPNAP1,
Rppn,
ropporin
252.3kb Sense Chr. 16 - 34649842 - 34678610 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108042 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663800 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12934100 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663818 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663795 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663798 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663792 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663797 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663808 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663803 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663801 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12961388 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663814 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663805 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12934111 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663794 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663789 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663810 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30663815 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104810497 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0