This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03954739_s1
See other 3110049I03RIK GE Assays ›
Gene Symbol:
3110049I03Rik
Gene Name:
RIKEN cDNA 3110049I03 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03954739_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 3110049I03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK014197.1 - 1 643 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream roundabout homolog 1 (Drosophila) Robo1 AW494633,
AW742721,
DUTT1,
Gm310
Sense Chr. 16 - 72027553 - 73047491 on NCBI Build GRCm38
Downstream RIKEN cDNA 8030451O07 gene 8030451O07Rik 109.3kb Anti-sense Chr. 16 - 72228720 - 72237867 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701440 AK088136.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701440 AK169558.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701440 NM_178666.5 refseq_mrna_mouse_NM
Whole Mouse Genome G4122A A_51_P390794 AK014197.1 mouse_transcriptome
Agilent Six_ChIP_03_of_03_v2 A_68_P30858136 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858154 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12583846 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858159 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858151 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858133 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858134 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858141 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858140 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12750349 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858157 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858138 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858143 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858156 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858146 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858147 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858144 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858135 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30858155 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1433172_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1433171_PM_at - -
Mouse Genome 430 2.0 Array 1433172_at - -
Mouse Genome 430 2.0 Array 1433171_at - -
Mouse Genome 430 2.0 Array 1433172_PM_at - -
Mouse Genome 430 2.0 Array 1433171_PM_at - -
Mouse Expression Set 430 1433172_at - -
Mouse Expression Set 430 1433172_PM_at - -
Mouse Expression Set 430 1433171_at - -
Mouse Expression Set 430 1433171_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433171_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433171_at - -
Affymetrix HT Mouse Genome 430B Array 1433172_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1433172_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0