This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03954008_s1
See other THAP2 GE Assays ›
Gene Symbol:
Thap2
Gene Name:
THAP domain containing, apoptosis associated protein 2
Gene Aliases:
2900040O07Rik, 9030625G08Rik, AI450385, AI649097
Chromosome Location:
Chr.10: 115369966 - 115384435 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03954008_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Thap2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK013630.1 - 1 447 80
AK080279.1 - 1721 80
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY557996.1 - 1 371 Chr. 10 - 115384002 - 115384443 [-] on Build GRCm38 80
GenBankmRNA AI838716.1 - 1 Chr. 10 - 115383917 - 115384242 [+] on Build GRCm38 80
GenBankmRNA BB229507.2 - 1 541 Chr. 10 - 115383918 - 115384590 [-] on Build GRCm38 80
GenBankmRNA BG072341.2 - 1 Chr. 10 - 115384002 - 115384425 [+] on Build GRCm38 80
GenBankmRNA BG079966.2 - 1 55 Chr. 10 - 115383449 - 115384127 [-] on Build GRCm38 80
GenBankmRNA BY712598.1 - 1 447 Chr. 10 - 115383978 - 115384516 [-] on Build GRCm38 80

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transmembrane protein 19 Tmem19 2810428F02Rik,
AW125171
-21.5kb Sense Chr. 10 - 115340739 - 115362524 on NCBI Build GRCm38
Downstream zinc finger,
C3H1-type containing
Zfc3h1 BC033596,
Ccdc131,
Psrc2
404.0 Anti-sense Chr. 10 - 115384920 - 115432772 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0