This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03953881_s1
See other 2900019E01RIK GE Assays ›
Gene Symbol:
2900019E01Rik
Gene Name:
RIKEN cDNA 2900019E01 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03953881_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2900019E01Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK013555.1 - 1 1343 94
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BE945320.1 - 1 Chr. 15 - 43532709 - 43533217 [-] on Build GRCm38 94
GenBankmRNA BF472917.1 - 1 205 Chr. 15 - 43532662 - 43533204 [+] on Build GRCm38 94

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ER membrane protein complex subunit 2 Emc2 4921531G14Rik,
AV060620,
AW209495,
Ttc35
-3.7kb Sense Chr. 15 - 43477214 - 43527777 on NCBI Build GRCm38
Downstream transmembrane protein 74 Tmem74 AA549547,
B230382K22Rik
333.5kb Anti-sense Chr. 15 - 43866695 - 43870612 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834000 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1430348_PM_at - -
Mouse Genome 430 2.0 Array 1430348_PM_at - -
Mouse Genome 430 2.0 Array 1430348_at - -
Mouse Expression Set 430 1430348_PM_at - -
Mouse Expression Set 430 1430348_at - -
Affymetrix HT Mouse Genome 430B Array 1430348_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430348_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0