This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03953533_s1
See other 2810410P21RIK GE Assays ›
Gene Symbol:
2810410P21Rik
Gene Name:
RIKEN cDNA 2810410P21 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03953533_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2810410P21Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK013077.1 - 1 843 87

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream pleckstrin homology domain interacting protein Phip 2810004D21Rik,
4632404O06Rik,
Ndrp,
Wdr11
-110.3kb Sense Chr. 9 - 82866159 - 82975700 on NCBI Build GRCm38
Downstream predicted gene 2065 Gm2065 10.1kb Anti-sense Chr. 9 - 83098137 - 83125776 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314073 - -
Whole Mouse Genome G4122A A_52_P793889 AK013077.1 mouse_transcriptome
Agilent Six_ChIP_02_of_03_v2 A_68_P26681775 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26681781 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26681783 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07566263 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26681777 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26681780 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26681779 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432908_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454412_PM_at - -
Mouse Genome 430 2.0 Array 1432908_PM_at - -
Mouse Genome 430 2.0 Array 1432908_at - -
Mouse Genome 430 2.0 Array 1454412_PM_at - -
Mouse Genome 430 2.0 Array 1454412_at - -
Mouse Expression Set 430 1432908_at - -
Mouse Expression Set 430 1454412_PM_at - -
Mouse Expression Set 430 1454412_at - -
Mouse Expression Set 430 1432908_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432908_at - -
Affymetrix HT Mouse Genome 430B Array 1454412_at - -
Affymetrix HT Mouse Genome 430B Array 1454412_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432908_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0