This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03952822_s1
See other 2610012C04RIK GE Assays ›
Gene Symbol:
2610012C04Rik
Gene Name:
RIKEN cDNA 2610012C04 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03952822_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2610012C04Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK011385.1 - 1 548 88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI120134.1 - 1 Chr. 1 - 135480568 - 135480966 [+] on Build GRCm38 87
GenBankmRNA BX526300.1 - 1 Chr. 1 - 135480560 - 135481079 [+] on Build GRCm38 88

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 1231 Mir1231 Gm23720,
mir-1231,
mmu-mir-1231
-25.7kb Sense Chr. 1 - 135454603 - 135454683 on NCBI Build GRCm38
Downstream predicted gene 4793 Gm4793 EG215714 103.9kb Anti-sense Chr. 1 - 135585351 - 135600699 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432606_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1432607_PM_at - -
Mouse Genome 430 2.0 Array 1432606_at - -
Mouse Genome 430 2.0 Array 1432607_PM_at - -
Mouse Genome 430 2.0 Array 1432606_PM_at - -
Mouse Genome 430 2.0 Array 1432607_at - -
Mouse Expression Set 430 1432606_at - -
Mouse Expression Set 430 1432606_PM_at - -
Mouse Expression Set 430 1432607_PM_at - -
Mouse Expression Set 430 1432607_at - -
Affymetrix HT Mouse Genome 430B Array 1432607_at - -
Affymetrix HT Mouse Genome 430B Array 1432606_at - -
Affymetrix HT Mouse Genome 430B Array 1432607_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432606_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0