This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03952680_s1
See other 2510017J16RIK GE Assays ›
Gene Symbol:
2510017J16Rik
Gene Name:
RIKEN cDNA 2510017J16 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03952680_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2510017J16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK010976.1 - 1 356 96
AK010977.1 - 1 356 96
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY710361.1 - 1 356 Chr. 5 - 3585112 - 3586053 [+] on Build GRCm38 96

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RNA binding motif protein 48 Rbm48 AW548102,
C030048B08Rik
Anti-sense Chr. 5 - 3583978 - 3596547 on NCBI Build GRCm38
Downstream peroxisomal biogenesis factor 1 Pex1 5430414H02Rik,
E330005K07Rik,
ZWS1
9.7kb Sense Chr. 5 - 3596066 - 3637230 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_51_P170894 AK010977.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P170894 AK010976.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1430393_PM_at - -
Mouse Genome 430 2.0 Array 1430393_at - -
Mouse Genome 430 2.0 Array 1430393_PM_at - -
Mouse Expression Set 430 1430393_PM_at - -
Mouse Expression Set 430 1430393_at - -
Affymetrix HT Mouse Genome 430B Array 1430393_at - -
Affymetrix HT Mouse Genome 430B Array 1430393_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 380502 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0