This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03951687_g1
See other IDI2 GE Assays ›
Gene Symbol:
Idi2
Gene Name:
isopentenyl-diphosphate delta isomerase 2
Gene Aliases:
4833405L16Rik, IPPI2
Chromosome Location:
Chr.13: 8952863 - 8960935 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03951687_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Idi2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177197.4 NP_796171.1 1 - 2 52 76
GenBank mRNA AK029364.1 - 1 - 2 53 76
AK081319.1 - 1 - 2 53 76
AY263178.1 - 1 - 2 53 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK029364.1, AK081319.1, AY263178.1, NM_177197.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Idi2

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cholesterol biosynthetic process
isopentenyl diphosphate metabolic process
isopentenyl-diphosphate delta-isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0