This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03949748_gH
See other PRDX1 GE Assays ›
Gene Symbol:
Prdx1
Gene Name:
peroxiredoxin 1
Gene Aliases:
MSP23, NkefA, OSF-3, OSF3, PAG, Paga, PrdxI, PrxI, TDX2, TPxA, Tdpx2, prx1
Chromosome Location:
Chr.4: 116685599 - 116700000 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03949748_gH
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prdx1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011034.4 NP_035164.1 1 - 2 60 77
GenBank mRNA AK002287.1 - 1 - 2 59 77
AK008711.1 - 1 - 2 62 77
AK010688.1 - 1 - 2 61 77
AK083243.1 - 1 - 2 61 77
AK145138.1 - 61 77
AK150797.1 - 1 - 2 61 77
AK151459.1 - 1 - 2 61 77
AK151760.1 - 1 - 2 59 77
AK163984.1 - 1 - 2 61 77
AK167624.1 - 1 - 2 61 77
AK169154.1 - 1 - 2 58 77
D16142.1 - 1 - 2 46 77
D21252.1 - 1 - 2 50 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK002287.1, AK008711.1, AK010688.1, AK083243.1, AK145138.1, AK150797.1, AK151459.1, AK151760.1, AK167624.1, AK169154.1, D16142.1, D21252.1, NM_011034.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prdx1

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to reactive oxygen species
response to oxidative stress
cell proliferation
removal of superoxide radicals
regulation of stress-activated MAPK cascade
erythrocyte homeostasis
natural killer cell mediated cytotoxicity
regulation of NF-kappaB import into nucleus
hydrogen peroxide catabolic process
oxidation-reduction process
peroxidase activity
protein binding
thioredoxin peroxidase activity
antioxidant activity
oxidoreductase activity
heme binding
identical protein binding
protein homodimerization activity
poly(A) RNA binding
peroxiredoxin activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0