This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03946522_m1
See other B930046M08RIK GE Assays ›
Gene Symbol:
B930046M08Rik
Gene Name:
RIKEN cDNA B930046M08 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03946522_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: B930046M08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK084781.1 - 3 - 4 612 70

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream family with sequence similarity 184,
member A
Fam184a 3110012E06Rik,
4930438C08Rik,
4930589M24Rik
Anti-sense Chr. 10 - 53633128 - 53751231 on NCBI Build GRCm38
Downstream mannosidase 1,
alpha
Man1a Man1a1,
PCR1
246.3kb Anti-sense Chr. 10 - 53904785 - 54076586 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

MicroArray Probes:

Agilent Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122828 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122828 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122828 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106290131 - -
MouseWG-6 v2 Expression BeadChip 100630605 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0