This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03944733_m1
See other A930015D03RIK GE Assays ›
Gene Symbol:
A930015D03Rik
Gene Name:
RIKEN cDNA A930015D03 gene
Gene Aliases:
-
Chromosome Location:
Chr.17: 35994504 - 36038174 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03944733_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: A930015D03Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_015618.2 1 - 2 55 104
GenBank mRNA AK020860.1 - 1 - 2 56 104
AK078380.1 - 1 - 2 106 104
AK142942.1 - 1 - 2 51 104
BC055918.1 - 1 - 2 33 104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY742198.1 - 1 - 2 56 Chr. 17 - 35994504 - 36038041 [+] on Build GRCm38 104
GenBankmRNA BY719272.1 - 1 - 2 106 Chr. 17 - 35994453 - 36038054 [+] on Build GRCm38 104
GenBankmRNA DT928325.1 - 1 - 2 33 Chr. 17 - 35994526 - 36037939 [+] on Build GRCm38 104
GenBankmRNA BB605749.2 - 1 - 2 51 Chr. 17 - 35994508 - 36038018 [+] on Build GRCm38 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream histocompatibility 2,
T region locus 24
H2-T24 H-2T24 Anti-sense Chr. 17 - 36003992 - 36020842 on NCBI Build GRCm38
Downstream predicted gene 6034 Gm6034 EG547347,
H2-gs14-2
4.6kb Sense Chr. 17 - 36042961 - 36058645 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000717.mm.1
This assay targets the same location as RTA transcript cluster(s) TC0400002975.rn

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127759 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115755 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110701 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700007387.mm.1 NR_015618.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700007387.mm.1 TC1700000717.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700007384.mm.1 NR_015618.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700007384.mm.1 TC1700000717.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 667698 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0