This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03941734_s1
See other 5830433D23RIK GE Assays ›
Gene Symbol:
5830433D23Rik
Gene Name:
RIKEN cDNA 5830433D23 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03941734_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5830433D23Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK017965.1 - 1 1256 91

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phosphatidylinositol transfer protein,
cytoplasmic 1
Pitpnc1 1110020B03Rik,
5830436L09Rik,
AI662802,
AI851387,
C330017I21Rik,
Dnr411,
RDGBB,
RDGBB1,
rdgB-beta
Sense Chr. 11 - 107207892 - 107470892 on NCBI Build GRCm38
Downstream predicted gene 11716 Gm11716 OTTMUSG00000003632 70.9kb Anti-sense Chr. 11 - 107335027 - 107344810 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432728_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454323_PM_at - -
Mouse Genome 430 2.0 Array 1454323_at - -
Mouse Genome 430 2.0 Array 1432728_at - -
Mouse Genome 430 2.0 Array 1432728_PM_at - -
Mouse Genome 430 2.0 Array 1454323_PM_at - -
Mouse Expression Set 430 1432728_PM_at - -
Mouse Expression Set 430 1454323_at - -
Mouse Expression Set 430 1454323_PM_at - -
Mouse Expression Set 430 1432728_at - -
Affymetrix HT Mouse Genome 430B Array 1432728_at - -
Affymetrix HT Mouse Genome 430B Array 1454323_at - -
Affymetrix HT Mouse Genome 430B Array 1454323_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432728_PM_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header