This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03941151_s1
See other 5730407M17RIK GE Assays ›
Gene Symbol:
5730407M17Rik
Gene Name:
RIKEN cDNA 5730407M17 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03941151_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 5730407M17Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK017517.1 - 1 547 64
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY603055.1 - 1 220 Chr. 2 - 34871435 - 34871826 [+] on Build GRCm38 64
GenBankmRNA BY605254.1 - 1 248 Chr. 2 - 34871407 - 34871826 [+] on Build GRCm38 64
GenBankmRNA BY610784.1 - 1 226 Chr. 2 - 34871429 - 34871826 [+] on Build GRCm38 64
GenBankmRNA BY717309.1 - 1 541 Chr. 2 - 34871113 - 34872049 [+] on Build GRCm38 64

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream proteasome (prosome,
macropain) 26S subunit,
non-ATPase,
5
Psmd5 1500032A03Rik,
AW475925,
S5b,
W91691,
mKIAA0072
-146.0 Anti-sense Chr. 2 - 34852089 - 34870962 on NCBI Build GRCm38
Downstream cutA divalent cation tolerance homolog-like Cutal D730039F16Rik 2.2kb Sense Chr. 2 - 34874365 - 34892133 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2304337 AI317200.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2304337 AK017517.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275992 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106030 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1431347_PM_at - -
Mouse Genome 430 2.0 Array 1431347_PM_at - -
Mouse Genome 430 2.0 Array 1431347_at - -
Mouse Expression Set 430 1431347_PM_at - -
Mouse Expression Set 430 1431347_at - -
Affymetrix HT Mouse Genome 430B Array 1431347_at - -
Affymetrix HT Mouse Genome 430B Array 1431347_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 101190242 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0