This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03939942_s1
See other 4933404M09RIK GE Assays ›
Gene Symbol:
4933404M09Rik
Gene Name:
RIKEN cDNA 4933404M09 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03939942_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4933404M09Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK016655.1 - 1 1749 62

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream chemokine (C-X-C motif) ligand 14 Cxcl14 1110031L23Rik,
1200006I23Rik,
AI414372,
BMAC,
BRAK,
KS1,
Kec,
MIP-2g,
MIP2gamma,
NJAC,
Scyb14,
bolekine
-40.9kb Sense Chr. 13 - 56288643 - 56296792 on NCBI Build GRCm38
Downstream predicted gene 10782 Gm10782 ENSMUSG00000074885 22.9kb Anti-sense Chr. 13 - 56362900 - 56368857 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2351255 AK016655.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2351255 AV278915.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4679 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060640 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10740460 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060617 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060662 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060663 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060658 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060621 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10700387 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060630 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060653 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10700388 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060604 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10658071 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060649 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060624 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060659 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060632 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060622 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10471065 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10528848 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060648 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060636 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060645 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060628 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10740455 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10482341 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P10482340 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060616 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060607 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060655 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060614 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060610 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060644 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060606 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060620 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P29060609 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1432664_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1454322_PM_at - -
Mouse Genome 430 2.0 Array 1454322_at - -
Mouse Genome 430 2.0 Array 1432664_PM_at - -
Mouse Genome 430 2.0 Array 1454322_PM_at - -
Mouse Genome 430 2.0 Array 1432664_at - -
Mouse Expression Set 430 1454322_PM_at - -
Mouse Expression Set 430 1432664_PM_at - -
Mouse Expression Set 430 1454322_at - -
Mouse Expression Set 430 1432664_at - -
Affymetrix HT Mouse Genome 430B Array 1454322_at - -
Affymetrix HT Mouse Genome 430B Array 1432664_at - -
Affymetrix HT Mouse Genome 430B Array 1454322_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1432664_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 817531 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106290463 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0