Assay Details
Gene Symbol: Gm11149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029465.1 | 4 - 5 | 537 | 70 | ||
GenBank mRNA | AK015955.1 | - | 4 - 5 | 538 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tetratricopeptide repeat domain 12 | Ttc12 | E330017O07Rik | -77.8kb | Anti-sense | Chr. 9 - 49436961 - 49486226 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 2310003N18 gene | 2310003N18Rik | 665.7kb | Anti-sense | Chr. 9 - 50232527 - 50233522 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
