This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03939223_m1
See other GM11149 GE Assays ›
Gene Symbol:
Gm11149
Gene Name:
predicted gene 11149
Gene Aliases:
ENSMUSG00000060603, ENSMUSG00000079564
Chromosome Location:
Chr.9: 49518288 - 49568025 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03939223_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gm11149
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_029465.1 1 - 2 75 64
GenBank mRNA AK015955.1 - 1 - 2 76 64
BC049643.1 - 1 - 2 45 64
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BU936741.1 - 1 - 2 48 Chr. 9 - 49518316 - 49562935 [+] on Build GRCm38 64
GenBankmRNA BY715520.1 - 1 - 2 76 Chr. 9 - 49518288 - 49567661 [+] on Build GRCm38 64

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tetratricopeptide repeat domain 12 Ttc12 E330017O07Rik -32.1kb Anti-sense Chr. 9 - 49436961 - 49486226 on NCBI Build GRCm38
Downstream RIKEN cDNA 2310003N18 gene 2310003N18Rik 712.3kb Anti-sense Chr. 9 - 50232527 - 50233522 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000594.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900004210.mm.1 NR_029465.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004210.mm.1 NR_029465.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004210.mm.1 TC0900000594.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900004211.mm.1 NR_029465.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004211.mm.1 NR_029465.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004211.mm.1 TC0900000594.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900004212.mm.1 NR_029465.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004212.mm.1 NR_029465.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900004212.mm.1 TC0900000594.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 421395 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0