This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03938799_m1
See other 4930458D05RIK GE Assays ›
Gene Symbol:
4930458D05Rik
Gene Name:
RIKEN cDNA 4930458D05 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03938799_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930458D05Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK015484.1 - 1 - 2 994 133
AK036594.1 - 1 - 2 2047 132
AK133807.1 - 1 - 2 983 132
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI413995.1 - 1 - 2 Chr. 5 - 100841628 - 100845475 [+] on Build GRCm38 132
GenBankmRNA BB028846.2 - 1 - 2 806 Chr. 5 - 100841632 - 100845859 [-] on Build GRCm38 132

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream 1-acylglycerol-3-phosphate O-acyltransferase 9 Agpat9 1-AGPAT,
1-AGPAT 9,
4933407I02Rik,
4933408F15,
9,
A230097K15Rik,
GPAT-3,
mGPAT3
Anti-sense Chr. 5 - 100845710 - 100899102 on NCBI Build GRCm38
Downstream predicted gene,
36793
Gm36793 63.9kb Sense Chr. 5 - 100910003 - 100913707 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2315012 AK036594.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2315012 AK015484.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199573 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144584 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290101 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99616 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136575 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2315012 AK036594.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2315012 AK015484.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136575 - -
Whole Mouse Genome G4122A A_52_P44111 AK036594.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P44111 AK015484.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108074 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136575 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453488_PM_at - -
Mouse Genome 430 2.0 Array 1453488_PM_at - -
Mouse Genome 430 2.0 Array 1454799_PM_at - -
Mouse Genome 430 2.0 Array 1453488_at - -
Mouse Genome 430 2.0 Array 1454799_at - -
Mouse Expression Set 430 1453488_at - -
Mouse Expression Set 430 1453488_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453488_at - -
Affymetrix HT Mouse Genome 430B Array 1453488_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 739333 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100430279 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0