This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03938055_s1
See other C130057N11RIK GE Assays ›
Gene Symbol:
C130057N11Rik
Gene Name:
RIKEN cDNA C130057N11 gene
Gene Aliases:
AI666698
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03938055_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C130057N11Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AI666698.1 - 1 99
AI845693.1 - 1 99
AK048403.1 - 1 3550 99
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CD541552.1 - 1 61 Chr. 2 - 154870701 - 154871258 [+] on Build GRCm38 99
GenBankmRNA AI467052.1 - 1 Chr. 2 - 154870690 - 154870949 [-] on Build GRCm38 99
GenBankmRNA BB371202.2 - 1 476 Chr. 2 - 154870297 - 154871010 [+] on Build GRCm38 99

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream hnRNP-associated with lethal yellow Raly AI663842,
Merc
Sense Chr. 2 - 154791097 - 154871010 on NCBI Build GRCm38
Downstream eukaryotic translation initiation factor 2,
subunit 2 (beta)
Eif2s2 2810026E11Rik,
38kDa,
AA408636,
AA571381,
AA986487,
AW822225,
D2Ertd303e,
EIF2,
EIF2B
400.0 Anti-sense Chr. 2 - 154871410 - 154892906 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent ABI Illumina

Array Name Probe IDs Targets Target Type
Whole Mouse Genome G4122A A_51_P515002 AK048403.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 423592 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102340736 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0