This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03931881_s1
See other 4833404L02RIK GE Assays ›
Gene Symbol:
4833404L02Rik
Gene Name:
RIKEN cDNA 4833404L02 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03931881_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4833404L02Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK029357.1 - 1 852 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AV248175.2 - 1 936 Chr. 7 - 17058566 - 17059423 [+] on Build GRCm38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream hypoxia inducible factor 3,
alpha subunit
Hif3a Ipas,
MOP7,
NEPAS,
bHLHe17
Anti-sense Chr. 7 - 17030993 - 17062427 on NCBI Build GRCm38
Downstream pregnancy specific glycoprotein 16 Psg16 Cea11,
PSG,
bCEA
14.6kb Sense Chr. 7 - 17074013 - 17133450 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

ABI Illumina

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 814228 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106110129 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0