This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03930422_g1
See other 2310003N18RIK GE Assays ›
Gene Symbol:
2310003N18Rik
Gene Name:
RIKEN cDNA 2310003N18 gene
Gene Aliases:
-
Chromosome Location:
Chr.9: 50232527 - 50233522 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03930422_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2310003N18Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK009126.1 - 1 - 2 134 81
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY708964.1 - 1 - 2 138 Chr. 9 - 50232523 - 50233182 [-] on Build GRCm38 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream neural cell adhesion molecule 1 Ncam1 CD56,
E-NCAM,
NCAM-1,
Ncam
-433.1kb Sense Chr. 9 - 49502134 - 49799427 on NCBI Build GRCm38
Downstream placenta expressed transcript 1,
opposite strand
Plet1os 2310014F07Rik 255.6kb Sense Chr. 9 - 50488799 - 50504805 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

MicroArray Probes:

Agilent

Array Name Probe IDs Targets Target Type
Agilent Six_ChIP_02_of_03_v2 A_68_P26502592 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502595 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502593 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502589 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07837176 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502588 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502597 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502583 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502580 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502600 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502582 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07766849 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502584 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07918934 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502594 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26502590 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0