This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03809346_m1
See other LATS2 GE Assays ›
Gene Symbol:
Lats2
Gene Name:
large tumor suppressor 2
Gene Aliases:
4932411G09Rik, AV277261, AW228608
Chromosome Location:
Chr.14: 57689662 - 57746123 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03809346_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lats2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153382.1 NP_700431.1 1 - 2 536 103
GenBank mRNA AY015061.1 - 1 - 2 418 103
BF147719.1 - 1 - 2 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002868.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
protein phosphorylation
cell cycle
mitotic nuclear division
hormone-mediated signaling pathway
phosphorylation
keratinocyte differentiation
cellular protein localization
hippo signaling
intracellular signal transduction
negative regulation of cyclin-dependent protein serine/threonine kinase activity
regulation of organ growth
cell division
negative regulation of canonical Wnt signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400016564.mm.1 NM_153382.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016564.mm.1 NM_153382.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016564.mm.1 TC1400002868.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0