This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03808664_g1
See other MPRIP GE Assays ›
Gene Symbol:
Mprip
Gene Name:
myosin phosphatase Rho interacting protein
Gene Aliases:
9530046C02, AA536749, AI647711, C76423, RIP3, Rhoip3, mKIAA0864, p116Rip
Chromosome Location:
Chr.11: 59662473 - 59780860 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03808664_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mprip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012027.2 NP_036157.2 2 - 3 530 86
NM_201245.3 NP_957697.1 2 - 3 530 86
GenBank mRNA AK044512.1 - 2 - 3 86
AK154807.1 - 2 - 3 537 86
AK155630.1 - 2 - 3 86
AK158296.1 - 2 - 3 519 86
AK170849.1 - 2 - 3 514 86
AK171089.1 - 2 - 3 519 86
BC039932.1 - 1 - 2 54 86
BC049803.1 - 2 - 3 170 86
BC063067.1 - 2 - 3 493 5721396 86
U73200.1 - 2 - 3 390 86

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mprip

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
maintenance of protein location in cell
negative regulation of myosin-light-chain-phosphatase activity
actin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0