This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03681969_m1
See other RHNO1 GE Assays ›
Gene Symbol:
Rhno1
Gene Name:
RAD9-HUS1-RAD1 interacting nuclear orphan 1
Gene Aliases:
2510047L19Rik, 5930416I19Rik
Chromosome Location:
Chr.6: 128357000 - 128362897 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03681969_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rhno1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027359.1 1 - 2 63 105
GenBank mRNA BE655013.1 - 1 - 2 63 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tubby-like protein 3 Tulp3 2310022L06Rik,
AI316887
-3.2kb Sense Chr. 6 - 128321161 - 128355872 on NCBI Build GRCm38
Downstream forkhead box M1 Foxm1 AA408308,
AW554517,
BB238854,
Fkh16,
Foxm1b,
HFH-11B,
MPHOSPH2,
Mpm2,
WIN
97.0 Anti-sense Chr. 6 - 128362994 - 128375886 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003201.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
cellular response to DNA damage stimulus
cell cycle
cellular response to ionizing radiation
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600024944.mm.1 TC0600003201.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600024958.mm.1 TC0600003201.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0