This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03646587_m1
See other SEC14L5 GE Assays ›
Gene Symbol:
Sec14l5
Gene Name:
SEC14-like 5 (S. cerevisiae)
Gene Aliases:
-
Chromosome Location:
Chr.16: 5146901 - 5187271 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03646587_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec14l5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127725.1 NP_001121197.1 12 - 13 1498 78
GenBank mRNA BC157907.1 - 12 - 13 1564 9088109 78
BC157990.1 - 12 - 13 1498 9007304 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000045.mm.1

Set Membership:

Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600000440.mm.1 NM_001127725.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000440.mm.1 NM_001127725.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000440.mm.1 TC1600000045.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600000439.mm.1 NM_001127725.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000439.mm.1 NM_001127725.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000439.mm.1 TC1600000045.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0