This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03455799_m1
See other 2010103J01RIK GE Assays ›
Gene Symbol:
2010103J01Rik
Gene Name:
RIKEN cDNA 2010103J01 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03455799_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 2010103J01Rik
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK008306.1 - LIT3439
2010103J01Rik
1 - 2 117 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RAB3 GTPase activating protein subunit 2 Rab3gap2 1110059F07Rik,
2010002H18Rik,
5830469C09,
AI851069,
AW743433,
RAB3-GAP150,
mKIAA0839
-25.4kb Sense Chr. 1 - 185204088 - 185286746 on NCBI Build GRCm38
Downstream bisphosphate 3'-nucleotidase 1 Bpnt1 BPntase 18.2kb Sense Chr. 1 - 185331965 - 185357769 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0