This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03048889_g1
See other DRG2 GE Assays ›
Gene Symbol:
Drg2
Gene Name:
developmentally regulated GTP binding protein 2
Gene Aliases:
AI255295, AI461605
Chromosome Location:
Chr.11: 60454617 - 60468724 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03048889_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Drg2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021354.1 NP_067329.1 4 - 5 435 90
GenBank mRNA AJ243590.1 - 4 - 5 435 90
AK012988.1 - 4 - 5 403 90
BC082564.1 - 4 - 5 391 30551418 90

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Drg2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
GTP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100005979.mm.1 NM_021354.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005979.mm.1 NM_021354.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100005979.mm.1 TC1100000754.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0