Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100006196.mm.1 |
NM_021349.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006196.mm.1 |
NM_021349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006196.mm.1 |
TC1100000774.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006193.mm.1 |
NM_021349.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006193.mm.1 |
NM_021349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006193.mm.1 |
TC1100000774.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006194.mm.1 |
NM_021349.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006194.mm.1 |
NM_021349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006194.mm.1 |
TC1100000774.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006195.mm.1 |
NM_021349.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006195.mm.1 |
NM_021349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006195.mm.1 |
TC1100000774.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006191.mm.1 |
TC1100000774.mm.1 |
MTA TC |