This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03024153_u1
See other GDF9 GE Assays ›
Gene Symbol:
Gdf9
Gene Name:
growth differentiation factor 9
Gene Aliases:
Gdf-9
Chromosome Location:
Chr.11: 53430878 - 53437903 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03024153_u1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gdf9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008110.2 NP_032136.2 1 469 133
GenBank mRNA AK136145.1 - 1 409 133
BC052667.1 - 1 399 30054181 133
L06444.1 - 1 377 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000604.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gdf9

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

oocyte growth
positive regulation of cell proliferation
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of cell growth
BMP signaling pathway
regulation of apoptotic process
regulation of MAPK cascade
cell development
SMAD protein signal transduction
regulation of progesterone secretion
cytokine activity
transforming growth factor beta receptor binding
growth factor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100004750.mm.1 NM_008110.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100004750.mm.1 NM_008110.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100004750.mm.1 TC1100000604.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100004751.mm.1 NM_008110.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100004751.mm.1 NM_008110.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100004751.mm.1 TC1100000604.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100004752.mm.1 NM_008110.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100004752.mm.1 NM_008110.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100004752.mm.1 TC1100000604.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0