This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm03012092_g1
See other FDPS GE Assays ›
Gene Symbol:
Fdps
Gene Name:
farnesyl diphosphate synthetase
Gene Aliases:
6030492I17Rik, AI256750, Fdpsl1, mKIAA1293
Chromosome Location:
Chr.3: 89093588 - 89101967 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm03012092_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fdps
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253751.1 NP_001240680.1 5 - 6 713 94
NM_134469.4 NP_608219.1 4 - 5 497 94
GenBank mRNA AF309508.1 - 3 - 4 402 94
AK088601.1 - 4 - 5 417 94
AK140881.1 - 4 - 5 489 94
AK166026.1 - 4 - 5 416 94
AK168008.1 - 4 - 5 416 94
BC048497.1 - 4 - 5 475 6531586 94
BC087886.1 - 4 - 5 420 6595030 94
CT010365.1 - 3 - 4 369 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fdps

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
steroid biosynthetic process
cholesterol biosynthetic process
steroid metabolic process
cholesterol metabolic process
isoprenoid biosynthetic process
sterol biosynthetic process
farnesyl diphosphate biosynthetic process
positive regulation of cholesterol biosynthetic process
positive regulation of cell growth involved in cardiac muscle cell development
dimethylallyltranstransferase activity
farnesyltranstransferase activity
geranyltranstransferase activity
transferase activity
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm