This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02620994_s1
See other MRGPRA7 GE Assays ›
Gene Symbol:
Mrgpra7
Gene Name:
MAS-related GPR, member A7
Gene Aliases:
MrgA7
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02620994_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mrgpra7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207535.1 NP_997418.1 1 246 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mrgpra7

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
biological_process
molecular_function
signal transducer activity
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004708 NM_207535.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004708 NM_207535.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10563727 - -
Affymetrix Mouse Gene 1.1 ST Array 10563725 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 105360050 - -
Illumina mouse-6 v1.0 expression beadchip 105360050 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1258944 - -
mouse-6 v1.1 (Illumina) GI_46430525-S - -
Illumina Sentrix mouse-6.v1 bead array 105360050 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 105360050 - -
MouseRef-8 v2 Expression BeadChip ILMN_2861051 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2861051 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0