This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02619667_s1
See other FPR3 GE Assays ›
Gene Symbol:
Fpr3
Gene Name:
formyl peptide receptor 3
Gene Aliases:
ALX, Fpr-rs1, Fpr-s1, Fprl1, LXA4-R, Lxa4r
Chromosome Location:
Chr.17: 17970458 - 17971677 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02619667_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fpr3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008042.2 NP_032068.2 1 1092 142
GenBank mRNA U78299.1 - 1140 142

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fpr3

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
signal transduction
G-protein coupled receptor signaling pathway
signal transducer activity
receptor activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700001733.mm.1 NM_008042.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700001733.mm.1 NM_008042.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700001733.mm.1 TC1700000214.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700001732.mm.1 NM_008042.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700001732.mm.1 NM_008042.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700001732.mm.1 TC1700000214.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0