Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400006873.mm.1 |
NM_001033773.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006873.mm.1 |
NM_001033773.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006873.mm.1 |
TC0400001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006870.mm.1 |
NM_001033773.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006870.mm.1 |
NM_001033773.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006870.mm.1 |
TC0400001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006875.mm.1 |
NM_001033773.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006875.mm.1 |
NM_001033773.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006875.mm.1 |
TC0400001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006874.mm.1 |
NM_001033773.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006874.mm.1 |
NM_001033773.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006874.mm.1 |
TC0400001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006871.mm.1 |
NM_001033773.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006871.mm.1 |
NM_001033773.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006871.mm.1 |
TC0400001017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400006872.mm.1 |
NM_001033773.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006872.mm.1 |
NM_001033773.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400006872.mm.1 |
TC0400001017.mm.1 |
MTA TC |