This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02601013_m1
See other SLC47A2 GE Assays ›
Gene Symbol:
Slc47a2
Gene Name:
solute carrier family 47, member 2
Gene Aliases:
4933429E10Rik
Chromosome Location:
Chr.11: 61301627 - 61342860 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02601013_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc47a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033542.2 NP_001028714.1 9 - 10 960 64
GenBank mRNA AK133432.1 - 9 - 10 961 64
BC147619.1 - 9 - 10 908 9056609 64
BC157913.1 - 9 - 10 908 9088130 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002908.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
drug transmembrane transport
biological_process
transmembrane transport
molecular_function
drug transmembrane transporter activity
antiporter activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27847846 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27847845 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27847847 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27847848 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965045 AK133432.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965045 NM_001033542.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350565 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896910 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566781 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623306 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334560 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925521 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276905 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443558 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57198 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 818396 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602585 AK041776.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602585 AK042297.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651249 AK144842.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651249 AK160894.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651249 BC017545.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651249 BC072577.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651249 BC079851.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651249 NM_145607.3 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611526 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946001 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434281 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10222 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99620 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730366 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3046 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141967 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647235 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630699 AK081095.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951849 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33177 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58575 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68453 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140896 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965045 AK133432.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965045 NM_001033542.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847837 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847851 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847840 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847841 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847846 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847833 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847844 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847849 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847848 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847835 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847845 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27847847 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02340579 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07364804 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07364851 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07364833 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07364879 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02340579 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07364833 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27847846 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27847847 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27847848 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27847845 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847783 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847818 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847835 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847802 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847774 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847721 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847685 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847709 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847846 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847828 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847778 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847689 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847848 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847627 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847635 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847670 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847674 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847754 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847823 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847728 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847655 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847769 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847736 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847659 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847651 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847724 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847791 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847766 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847799 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847730 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847735 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847682 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847708 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847815 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847632 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847779 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847849 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847810 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847797 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847707 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847723 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847812 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847692 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847739 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847679 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847726 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847658 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847768 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847776 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847704 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847748 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847691 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847745 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847733 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847793 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847820 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847847 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847697 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847630 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847834 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847702 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847844 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847809 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847813 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847851 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847752 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847782 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847790 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847831 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847777 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847641 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847770 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847794 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847716 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847672 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847788 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847800 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847720 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27847772 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847820 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847818 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847833 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847829 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847849 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09491810 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847851 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08984374 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847810 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09517360 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847830 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09229654 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09551617 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847840 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09551616 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847835 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847841 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847815 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847826 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847812 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09491636 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847837 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847825 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09517359 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09551605 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847809 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847824 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27847834 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10386671 - -
Affymetrix Mouse Gene 1.0 ST Array 10386670 - -
Affymetrix Mouse Gene 1.0 ST Array 10386669 - -
Affymetrix Mouse Gene 1.0 ST Array 10386676 - -
Affymetrix Mouse Gene 1.0 ST Array 10386681 - -
Affymetrix Mouse Gene 1.0 ST Array 10386678 - -
Affymetrix Mouse Gene 1.0 ST Array 10386666 - -
Affymetrix Mouse Gene 1.0 ST Array 10386672 - -
Affymetrix Mouse Gene 1.0 ST Array 10386673 - -
Affymetrix Mouse Gene 1.0 ST Array 10386667 - -
Affymetrix Mouse Gene 1.0 ST Array 10386680 - -
Affymetrix Mouse Gene 1.0 ST Array 10386679 - -
Affymetrix Mouse Gene 1.0 ST Array 10386682 - -
Affymetrix Mouse Gene 1.0 ST Array 10386668 - -
Affymetrix Mouse Gene 1.0 ST Array 10386674 - -
Affymetrix Mouse Gene 1.0 ST Array 10386677 - -
Affymetrix Mouse Gene 1.0 ST Array 10386675 - -
Affymetrix Mouse Gene 1.1 ST Array 10386665 - -
Affymetrix Mouse Clariom S Array PSR1100026374.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026374.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026374.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026376.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026376.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026376.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026389.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026389.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026389.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026383.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026383.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026383.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026372.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026372.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026372.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026387.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026387.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026387.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026382.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026382.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026382.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026392.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026392.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026392.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026373.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026373.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026373.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026386.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026386.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026386.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026379.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026379.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026379.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026385.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026385.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026385.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026378.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026378.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026378.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026388.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026388.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026388.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026381.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026381.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026381.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026375.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026375.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026375.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026380.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026380.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026380.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026377.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026377.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026377.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026384.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026384.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026384.mm.1 TC1100002908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100026391.mm.1 NM_001033542.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100026391.mm.1 NM_001033542.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100026391.mm.1 TC1100002908.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.1 expression beadchip ILMN_1242117 - -
MouseRef-8 v2 Expression BeadChip ILMN_3082420 - -
MouseRef-8 v2 Expression BeadChip ILMN_3011926 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3082420 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3011926 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0