This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02529550_s1
See other UBE2DNL1 GE Assays ›
Gene Symbol:
Ube2dnl1
Gene Name:
ubiquitin-conjugating enzyme E2D N-terminal like 1
Gene Aliases:
Ube2dnl
Chromosome Location:
Chr.X: 114905012 - 114905941 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02529550_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2dnl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276396.1 NP_001263325.1 2 668 135
GenBank mRNA BC048523.1 - 2 668 135

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream kelch-like 4 Klhl4 B130014A11,
C130018J01Rik
-344.3kb Sense Chr. X - 114474333 - 114561129 on NCBI Build GRCm38
Downstream ubiquitin-conjugating enzyme E2D N-terminal like 2 Ube2dnl2 4930524E20Rik 1.6kb Sense Chr. X - 114907582 - 114908510 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075549 NR_003645.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075549 BC048523.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836571 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075549 NR_003645.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075549 BC048523.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P638041 NR_003645.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P638041 BC048523.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10601532 - -
Affymetrix Mouse Gene 1.0 ST Array 10601533 - -
Affymetrix Mouse Gene 1.1 ST Array 10601531 - -
Affymetrix Mouse Clariom S Array PSR0X00007468.mm.1 NM_001276396.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007468.mm.1 TC0X00001146.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00007467.mm.1 NM_001276396.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007467.mm.1 TC0X00001146.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2739355 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2739355 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2779647 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0