This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02344096_g1
See other ARHGAP33OS GE Assays ›
Gene Symbol:
Arhgap33os
Gene Name:
Rho GTPase activating protein 33, opposite strand
Gene Aliases:
Gm1082
Chromosome Location:
Chr.7: 30515122 - 30522193 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02344096_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap33os
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036630.1 11 - 12 1750 162
GenBank mRNA BC065044.1 - 11 - 12 1749 5685982 162

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700004980.mm.1 NR_036630.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004980.mm.1 NR_036630.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004980.mm.1 TC0700000565.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700004981.mm.1 NR_036630.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004981.mm.1 NR_036630.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004981.mm.1 TC0700000565.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0