This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02343997_m1
See other SRFBP1 GE Assays ›
Gene Symbol:
Srfbp1
Gene Name:
serum response factor binding protein 1
Gene Aliases:
2810036K01Rik, p49/STRAP
Chromosome Location:
Chr.18: 52465672 - 52490738 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02343997_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srfbp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026040.3 NP_080316.3 1 - 2 97 85
GenBank mRNA AK012865.1 - 1 - 2 99 85
AK141882.1 - 91 85
AK144845.1 - 96 85
AK147692.1 - 75 85
AY611629.1 - 1 - 2 70 85
BC024338.1 - 74 5009612 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800000523.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Srfbp1

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
maturation of SSU-rRNA
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800003789.mm.1 NM_026040.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800003789.mm.1 NM_026040.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800003789.mm.1 TC1800000523.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800003790.mm.1 NM_026040.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800003790.mm.1 NM_026040.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800003790.mm.1 TC1800000523.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0