This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02342766_g1
See other ACTR3 GE Assays ›
Gene Symbol:
Actr3
Gene Name:
ARP3 actin-related protein 3
Gene Aliases:
1200003A09Rik, Arp3
Chromosome Location:
Chr.1: 125392905 - 125435727 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02342766_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Actr3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205385.1 NP_001192314.1 4 - 5 435 138
NM_001205386.1 NP_001192315.1 4 - 5 432 138
NM_023735.2 NP_076224.1 3 - 4 627 138
GenBank mRNA AK004554.1 - 3 - 4 441 138
AK083343.1 - 3 - 4 445 138
AK145536.1 - 3 - 4 447 138
AK147931.1 - 3 - 4 277 138
AK150487.1 - 3 - 4 329 138
AK150589.1 - 3 - 4 405 138
AK150991.1 - 3 - 4 385 138
AK152865.1 - 3 - 4 422 138
AK153079.1 - 3 - 4 448 138
AK153150.1 - 3 - 4 422 138
AK153204.1 - 3 - 4 328 138
AK159135.1 - 3 - 4 414 138
AK159668.1 - 3 - 4 339 138
AK159775.1 - 3 - 4 426 138
AK160047.1 - 3 - 4 287 138
AK166529.1 - 4 - 5 379 138
AK166586.1 - 4 - 5 379 138
AK167148.1 - 3 - 4 386 138
AK167891.1 - 3 - 4 384 138
AK168568.1 - 3 - 4 314 138
AK168771.1 - 3 - 4 400 138
BC005557.1 - 3 - 4 390 3156748 138
BC080806.1 - 4 - 5 393 5350752 138
BC108386.1 - 3 - 4 358 5051125 138

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Actr3

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
establishment or maintenance of cell polarity
asymmetric cell division
positive regulation of lamellipodium assembly
meiotic chromosome movement towards spindle pole
cell projection organization
positive regulation of actin filament polymerization
meiotic cytokinesis
Arp2/3 complex-mediated actin nucleation
regulation of myosin II filament organization
positive regulation of neuron differentiation
positive regulation of dendrite morphogenesis
meiotic cell cycle
positive regulation of filopodium assembly
spindle localization
cilium morphogenesis
nucleotide binding
actin binding
structural constituent of cytoskeleton
protein binding
ATP binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100024046.mm.1 NM_001205385.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024046.mm.1 NM_001205385.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024046.mm.1 NM_001205386.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024046.mm.1 NM_001205386.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024046.mm.1 NM_023735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024046.mm.1 NM_023735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024046.mm.1 TC0100002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024050.mm.1 NM_023735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024050.mm.1 NM_023735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024050.mm.1 TC0100002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024053.mm.1 NM_001205385.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024053.mm.1 NM_001205385.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024053.mm.1 NM_001205386.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024053.mm.1 NM_001205386.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024053.mm.1 NM_023735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024053.mm.1 NM_023735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024053.mm.1 TC0100002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024054.mm.1 NM_001205385.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024054.mm.1 NM_001205385.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024054.mm.1 NM_001205386.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024054.mm.1 NM_001205386.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024054.mm.1 NM_023735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024054.mm.1 NM_023735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024054.mm.1 TC0100002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024049.mm.1 NM_001205385.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024049.mm.1 NM_001205385.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024049.mm.1 NM_001205386.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024049.mm.1 NM_001205386.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024049.mm.1 NM_023735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024049.mm.1 NM_023735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024049.mm.1 TC0100002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024045.mm.1 NM_001205385.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024045.mm.1 NM_001205385.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024045.mm.1 NM_001205386.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024045.mm.1 NM_001205386.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024045.mm.1 NM_023735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024045.mm.1 NM_023735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024045.mm.1 TC0100002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024051.mm.1 NM_023735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024051.mm.1 NM_023735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024051.mm.1 TC0100002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024048.mm.1 NM_001205385.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024048.mm.1 NM_001205385.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024048.mm.1 NM_001205386.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024048.mm.1 NM_001205386.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024048.mm.1 NM_023735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024048.mm.1 NM_023735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024048.mm.1 TC0100002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100024047.mm.1 NM_001205385.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024047.mm.1 NM_001205385.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024047.mm.1 NM_001205386.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024047.mm.1 NM_001205386.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024047.mm.1 NM_023735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100024047.mm.1 NM_023735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100024047.mm.1 TC0100002993.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0