This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02029837_s1
See other ZFP950 GE Assays ›
Gene Symbol:
Zfp950
Gene Name:
zinc finger prtoein 950
Gene Aliases:
5830428H23Rik, AU016073, BC055757, Gm34518, Zfp826, mszf84
Chromosome Location:
Chr.19: 61051147 - 61313507 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02029837_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp950
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK047931.1 - 1 85 134
BC029127.1 - 1 880 134
BC040780.1 - 1 880 134

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

DNA-binding transcription factor zinc finger transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P135534 BC086681.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P135534 AK047931.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P135534 AK139999.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P135534 AK039536.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P135534 BC029127.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186122 AK047931.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186122 BC029127.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186122 BC040780.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186122 AK047931.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186122 BC029127.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2186122 BC040780.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01503957 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06175151 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06175186 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06175168 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159398 - -
Whole Mouse Genome G4122A A_52_P681348 BC029127.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P681348 BC040780.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P96087 AB010342.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P282935 - -
Whole Mouse Genome G4122A A_51_P486251 BC029127.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P486251 BC040780.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06175186 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159398 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01503957 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P32229035 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10468888 - -
Affymetrix Mouse Gene 1.1 ST Array 10468885 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 807235 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.1 expression beadchip ILMN_2727146 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0