This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm02018719_g1
See other GOLGA7 GE Assays ›
Gene Symbol:
Golga7
Gene Name:
golgi autoantigen, golgin subfamily a, 7
Gene Aliases:
AB041568, C130038N16Rik, GCP16, GOLGA3AP1, HSPC041, R75586
Chromosome Location:
Chr.8: 23241326 - 23257183 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm02018719_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Golga7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042484.1 NP_001035949.1 6 1138 99
NM_020585.2 NP_065610.2 5 1351 99
GenBank mRNA AB041568.1 - 5 1287 99
AK048175.1 - 6 1114 99
AK077582.1 - 6 1112 99
AK150462.1 - 6 1132 99
AK150696.1 - 5 1293 99
AK151353.1 - 6 1132 99
AK152520.1 - 6 1132 99
AK152606.1 - 6 1111 99
AK153213.1 - 6 1132 99
AK158227.1 - 5 1257 99
AK170129.1 - 6 1117 99
AK211526.1 - 1 168 99
AK212515.1 - 1 168 99
BC016894.1 - 6 1090 4223937 99
BC019697.1 - 5 1276 99
BC058523.1 - 5 1194 5717934 99
BC093518.1 - 6 1108 3157520 99
BC103796.1 - 6 1103 6519018 99

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Golga7

Gene Ontology Categories:

Function(s) Process(es)

Golgi to plasma membrane transport
peptidyl-L-cysteine S-palmitoylation
Golgi to plasma membrane protein transport
protein stabilization
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25715745 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25715748 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25715749 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25715751 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2120682 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027323 BC103796.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027323 BC019697.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027323 BC058523.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027323 BC093518.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027323 AK158227.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42625 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 871819 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115248 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929083 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 816519 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742390 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53863 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626490 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303475 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166602 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65330 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174903 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110767 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2120682 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027323 BC103796.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027323 BC019697.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027323 BC058523.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027323 BC093518.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027323 AK158227.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715741 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715740 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715732 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715733 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715745 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715769 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715755 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715764 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715758 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715749 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715737 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715736 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715766 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715762 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715731 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715748 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715729 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715738 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715760 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715768 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715757 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715742 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715752 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25715727 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174903 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110767 - -
Whole Mouse Genome G4122A A_51_P478942 BC103796.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P478942 BC019697.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P478942 BC058523.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P478942 BC093518.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P478942 AK158227.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P398018 AK089856.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130901 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174903 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110767 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06943719 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06943713 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06943710 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06943715 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06943717 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25715749 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25715751 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25715745 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25715748 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715733 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715777 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715576 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715710 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715787 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715693 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715781 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715703 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715696 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715738 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715679 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715641 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715766 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715772 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715662 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715677 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715697 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715749 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715671 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715741 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715578 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715655 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715695 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715692 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715776 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715691 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715779 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715686 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715716 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715676 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715786 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715593 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715600 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715669 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715714 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715585 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715651 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715575 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715736 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715732 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715731 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715755 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715660 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715680 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715690 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715582 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715646 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715584 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715742 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715581 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715769 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715715 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715708 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715729 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715773 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715748 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715718 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715752 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715596 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715608 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715705 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715727 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25715640 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715701 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07306074 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715768 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715752 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07317787 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715772 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715725 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715647 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715762 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715738 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715654 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715766 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06943717 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715729 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715695 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07339202 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06943710 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07085500 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715765 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715709 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715757 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715652 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715777 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06842984 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715776 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715733 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06954907 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715742 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715710 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715748 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715697 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715705 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07133824 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715779 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07420354 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715741 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715773 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07420355 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715640 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715714 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715643 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715769 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715749 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715649 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715712 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715755 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715637 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715655 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715775 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715719 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715703 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715708 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715727 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07373722 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715644 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715641 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715782 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715696 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07331195 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715718 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715716 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25715715 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06768719 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10577638 - -
Affymetrix Mouse Gene 1.0 ST Array 10577635 - -
Affymetrix Mouse Gene 1.0 ST Array 10577639 - -
Affymetrix Mouse Gene 1.0 ST Array 10532309 - -
Affymetrix Mouse Gene 1.0 ST Array 10577640 - -
Affymetrix Mouse Gene 1.0 ST Array 10577634 - -
Affymetrix Mouse Gene 1.0 ST Array 10577636 - -
Affymetrix Mouse Gene 1.0 ST Array 10577637 - -
Affymetrix HT MG-430 PM Array Plate 1415672_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10577633 - -
Affymetrix Mouse Gene 1.1 ST Array 10570955 - -
Affymetrix Mouse Gene 1.1 ST Array 10532308 - -
Mouse Genome 430 2.0 Array 1415672_at - -
Mouse Genome 430 2.0 Array 1415672_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 57437_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 57437_at - -
Mouse Expression Set 430 1415672_PM_at - -
Mouse Expression Set 430 1415672_at - -
Mouse Expression Set 430 1415672_PM_at - -
Mouse Expression Set 430 1415672_at - -
Murine Genome U74v2 Set 93256_at - -
Murine Genome U74v2 Set 93256_PM_at - -
Mouse Genome 430A 2.0 Array 1415672_at - -
Mouse Genome 430A 2.0 Array 1415672_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1415672_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1415672_at - -
Affymetrix HT Mouse Genome 430B Array 1415672_at - -
Affymetrix HT Mouse Genome 430B Array 1415672_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 898929 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 106760687 - -
MouseWG-6 v2 Expression BeadChip 6980121 - -
Illumina mouse-6 v1.0 expression beadchip 106760687 - -
Illumina mouse-6 v1.0 expression beadchip 6980121 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006980121 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2666519 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2666519 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1254148 - -
mouse-6 v1.1 (Illumina) scl057437.1_41-S - -
Illumina Sentrix mouse-6.v1 bead array 6980121 - -
Illumina Sentrix mouse-6.v1 bead array 106760687 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl057437.1_41-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980121 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 106760687 - -
MouseRef-8 v2 Expression BeadChip ILMN_2666519 - -
MouseRef-8 v2 Expression BeadChip ILMN_3157764 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254148 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3157764 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2666519 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0