Assay Details
Gene Symbol: 1110019D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045995.1 | 3 | 1307 | 125 | ||
GenBank mRNA | AK003812.1 | - | 3 | 1307 | 125 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 2610001J05 gene | 2610001J05Rik | C79299 | -23.6kb | Anti-sense | Chr. 6 - 13869074 - 13871483 on NCBI Build GRCm38 |
Downstream |
protein phosphatase 1, regulatory (inhibitor) subunit 3A |
Ppp1r3a |
GM, RG1, RGL |
817.4kb | Anti-sense | Chr. 6 - 14713822 - 14755274 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2240468 | AK003812.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2240468 | AW048707.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745188 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554122 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798130 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228265 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826371 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409585 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 709987 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674543 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580115 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84854 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181907 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955288 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360052 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890897 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732197 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 845905 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367986 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 586688 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 653836 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80002 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184717 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593153 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 126419 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373553 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878726 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494409 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348957 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19090 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275496 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394280 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221910 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950703 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168324 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402069 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363952 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284316 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482132 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 598285 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 557801 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417392 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508331 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747944 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 344414 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282321 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523140 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717185 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 173241 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236327 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 956118 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539020 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279534 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85980 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960479 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633708 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181445 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936793 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138052 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405124 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266135 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 323434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 479420 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704962 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94906 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84883 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39880 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 20166 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 143981 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 25284 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47855 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156105 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57767 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56616 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1925 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178992 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134658 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815574 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815319 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01268898 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815545 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815506 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815588 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P134658 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815428 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815479 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178992 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815388 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815340 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815373 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815308 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05815355 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01268820 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102539 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05815506 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05815373 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05815319 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178992 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05815545 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P134658 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249360 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P05487871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249348 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249399 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249400 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P05331264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249343 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249381 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249345 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249374 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249366 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P05342953 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249401 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P05002379 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249379 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249330 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249358 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249342 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249394 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249393 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P05212717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249335 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249321 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P05143732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P05342955 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P05002386 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249318 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P05342960 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249398 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249328 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24249369 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1453434_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1453434_at | - | - |
Mouse Genome 430 2.0 Array | 1453434_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600000662.mm.1 | NR_045995.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600000662.mm.1 | NR_045995.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600000662.mm.1 | TC0600000099.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600000660.mm.1 | NR_045995.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600000660.mm.1 | NR_045995.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600000660.mm.1 | TC0600000099.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600000661.mm.1 | NR_045995.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600000661.mm.1 | NR_045995.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600000661.mm.1 | TC0600000099.mm.1 | MTA TC |
Mouse Expression Set 430 | 1453434_at | - | - |
Mouse Expression Set 430 | 1453434_PM_at | - | - |
Murine Genome U74v2 Set | 140189_at | - | - |
Murine Genome U74v2 Set | 140189_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453434_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1453434_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 597644 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 101170113 | - | - |
