Assay Details
Gene Symbol: Gjb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK134742.1 | - | 1 | 466 | 65 | |
BC026833.1 | - | 2 | 638 | 3483365 | 65 | |
X84215.1 | - | 2 | 673 | 65 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32563826 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32563828 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32563827 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32563825 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096967 | BC026833.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096967 | NM_008124.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2096967 | AK134742.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973522 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 178941 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894622 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880013 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99410 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351766 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930380 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139555 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104046 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096967 | BC026833.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096967 | NM_008124.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2096967 | AK134742.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P462422 | M63802.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P462422 | X84215.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P462422 | BC026833.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P462422 | NM_008124.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P462422 | AK134742.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563789 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563778 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563750 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563790 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563756 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563779 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563771 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563760 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563768 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563758 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563748 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563776 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563787 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563753 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563781 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563792 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563782 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563759 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563784 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563770 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563780 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563775 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563773 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32563786 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139555 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104046 | - | - |
Whole Mouse Genome G4122A | A_51_P462422 | M63802.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P462422 | X84215.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P462422 | BC026833.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P462422 | NM_008124.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P462422 | AK134742.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114595 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104046 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139555 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15104824 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15104834 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15104823 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15104835 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P15104822 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32563825 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32563826 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32563827 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P32563828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563846 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563719 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563794 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32563813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563746 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14721702 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563787 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563812 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563775 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14721694 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563748 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563776 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14931691 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15104837 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563806 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563825 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15104825 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563740 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563792 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14806599 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14890750 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563850 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14931680 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563768 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14721700 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563751 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563778 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563743 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14918416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14695959 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15116997 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14931665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563784 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14926566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563791 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14621505 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14782182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563803 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563770 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563805 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14710810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15028641 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563724 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14513641 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14721696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15104827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563786 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563779 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563819 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563820 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14890747 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563773 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563762 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563814 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563809 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14926568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563758 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563797 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563729 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563726 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32563829 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10601162 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10601163 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448766_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448767_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10601161 | - | - |
Mouse Genome 430 2.0 Array | 1448766_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1448766_at | - | - |
Mouse Genome 430 2.0 Array | 1448767_s_at | - | - |
Mouse Genome 430 2.0 Array | 1448767_PM_s_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14618_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14618_PM_at | - | - |
Murine Genome U74v2 Set | 93771_at | - | - |
Murine Genome U74v2 Set | 93771_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448766_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448767_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448767_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448766_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 1240441 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001240441 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219151 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1219151 | - | - |
mouse-6 v1.1 (Illumina) | scl54743.5_163-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1240441 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl54743.5_163-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1240441 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2948476 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2948476 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl54743.5_163-S | - | - |
