This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01737888_m1
See other ALDH1L1 GE Assays ›
Gene Symbol:
Aldh1l1
Gene Name:
aldehyde dehydrogenase 1 family, member L1
Gene Aliases:
1810048F20Rik, FDH, Fthfd, Neut2
Chromosome Location:
Chr.6: 90550767 - 90599171 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01737888_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027406.1 NP_081682.1 6 - 7 866 88
GenBank mRNA BC022112.1 - 6 - 7 866 88
BC024055.1 - 6 - 7 823 5098789 88
BC025939.1 - 6 - 7 848 5099360 88
BC028817.1 - 6 - 7 813 5101815 88
BC030722.1 - 6 - 7 850 5098671 88
BC030723.1 - 6 - 7 782 5100007 88
BC030727.1 - 6 - 7 852 5099026 88
BC030730.1 - 6 - 7 866 5101835 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001076.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldh1l1

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

one-carbon metabolic process
metabolic process
biosynthetic process
10-formyltetrahydrofolate catabolic process
oxidation-reduction process
catalytic activity
aldehyde dehydrogenase (NAD) activity
aldehyde dehydrogenase [NAD(P)+] activity
protein binding
methyltransferase activity
formyltetrahydrofolate dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
hydroxymethyl-, formyl- and related transferase activity
protein complex binding
aldehyde dehydrogenase (NADP+) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0