This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01722563_m1
See other NUDT6 GE Assays ›
Gene Symbol:
Nudt6
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 6
Gene Aliases:
Asfgf2b
Chromosome Location:
Chr.3: 37404982 - 37420186 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01722563_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291044.1 NP_001277973.1 4 - 5 570 85
GenBank mRNA AF453427.1 - 4 - 5 679 85
AF453428.1 - 4 - 5 556 85
AK145299.1 - 4 - 5 562 85
BC146000.1 - 4 - 5 564 40131303 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001871.mm.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

nucleotide phosphatase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
thiamine-pyrophosphatase activity
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
UDP-2,3-diacylglucosamine hydrolase activity
bis(5'-nucleosyl)-tetraphosphatase activity
dATP pyrophosphohydrolase activity
pyrophosphatase activity
hydrolase activity
dihydroneopterin monophosphate phosphatase activity
dihydroneopterin triphosphate pyrophosphohydrolase activity
dITP diphosphatase activity
dTTP diphosphatase activity
XTP diphosphatase activity
ATP-dependent 5'-3' DNA helicase activity
phosphocholine hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300014487.mm.1 NM_153561.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300014487.mm.1 TC0300001871.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0