This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01719524_m1
See other CRAMP1L GE Assays ›
Gene Symbol:
Cramp1l
Gene Name:
Crm, cramped-like (Drosophila)
Gene Aliases:
5830477H08Rik, AI256853, AI790734, Tce4, mKIAA1426
Chromosome Location:
Chr.17: 24961226 - 25015846 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01719524_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cramp1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020608.3 NP_065633.2 16 - 17 3184 86
GenBank mRNA AK089393.1 - 16 - 17 3444 86
AK129356.1 - 7 - 8 1360 86
AK147411.1 - 16 - 17 3185 86
AK147449.1 - 16 - 17 3185 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001650.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cramp1l

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
DNA binding
chromatin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700014892.mm.1 NM_020608.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700014892.mm.1 NM_020608.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700014892.mm.1 TC1700001650.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0