This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01718956_m1
See other IGHM GE Assays ›
Gene Symbol:
Ighm
Gene Name:
immunoglobulin heavy constant mu
Gene Aliases:
AI326478, Igh-6, Igh-M, Igh6, Igm, muH
Chromosome Location:
Chr.12: 113418826 - 113422730 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01718956_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ighm
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK088666.1 - 1515 109
BC018315.1 - 1744 109
BC053409.1 - 1764 109
V00821.1 - 1 - 2 78 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0